Documents
CLS7160
CellSTACK® Chamber, 10 STACK
About This Item
UNSPSC Code:
41122104
sterility
sterile
manufacturer/tradename
Corning
availability
available only in China
Related Categories
General description
The CellSTACK® Chamber is a sterile and nonpyrogenic cell culture vessel. They consist of ten chambers, having a total cell growth area of 6360 cm2.
Application
CellSTACK® Chambers have been used extensively in large-scale cell culture experiments, aiding in the attachment and growth of diverse kinds of cells.
Features and Benefits
  • The TM="CellSTACK"] Chambers possess superior mechanical strength and structural integrity.
  • The presence of self-venting caps helps in the gas exchange process.
  • These are 100% leak evaluated.
  • The CellSTACK Chambers have larger openings with threaded closures, ensuring easy accessibility to cells while maintaining a secure seal to prevent contamination.
  • The CellSTACK chamber has a sterility assurance level of 10-6.
  • A hydrophilic and negatively charged surface enhances cell growth and attachment.
  • A minimum of 95% confluency is necessary for optimum cell growth and attachment
Legal Information
CellSTACK is a registered trademark of Corning, Inc.
Visit the Document Library
Residues W320 and Y328 within the binding site of the m-opioid receptor influence opiate ligand bias
J. Daniel Hothersall
Neuropharmacology (2017)
AAV-Dominant Negative Tumor Necrosis Factor (DN-TNF) Gene Transfer to the Striatum Does Not Rescue Medium Spiny Neurons in the YAC128 Mouse Model of Huntington's Disease
Alto LT, et al.
PLoS ONE (2014)
Upstream process optimization and micro- and macrocarrier screening for large-scale production of the oncolytic H-1 protoparvovirus
Wohlfarth D, et al.
Applied Microbiology and Biotechnology (2021)