Key Documents
CLS9903
Corning® CellSTACK® cell culture chambers
10 chambers, TC-treated, universal cap, sterile, 4/cs
Synonym(s):
Corning container
Select a Size
1 EA
$701.00
4 EA
$2,150.00
$701.00
Check Cart for Availability
Select a Size
Change View
1 EA
$701.00
4 EA
$2,150.00
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
$701.00
Check Cart for Availability
material
cap (universal)
sterility
sterile
packaging
pack of 1 ea
case of 4 ea
manufacturer/tradename
Corning 9903
suitability
suitable for (scale-up cell culture)
General description
  • Ideal for cell culture scale-up
  • Culture chambers produce cell yields in the 107 - 109 range
  • Useful alternatives to multiple roller bottles or spinner flasks.
  • High mechanical strength and structural integrity
  • 100% leak tested prior to shipping
  • Certified nonpyrogenic and sterilized by gamma irradiation, with low particulate contamination from modern manufacturing methods
  • Easy to use larger openings with threaded closures and vented caps
  • Large 26 mm wide filling ports allow direct access to chamber bottom, with more options for sterile transfer of media and cultures
Legal Information
CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)