Key Documents
CLS6620
Corning® Step-R Repeating Pipettor Syringe Tips
volume (0.05 mL), non-sterile
Synonym(s):
pipet tips
Select a Size
100 EA
$332.00
$332.00
Estimated to ship on14 May 2025
Select a Size
Change View
100 EA
$332.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
(0.05 mL)
sterility:
non-sterile
packaging:
pack of 100 ea
manufacturer/tradename:
Corning 6620
$332.00
Estimated to ship on14 May 2025
sterility
non-sterile
packaging
pack of 100 ea
manufacturer/tradename
Corning 6620
volume
(0.05 mL)
General description
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)