Key Documents
BR28205
BRAND® cotton roving
degreases cotton wool, density - Approx 1.3g per m
Synonym(s):
BLAUBRAND®, ETERNA, Graduated Pipettes, Pipettes
Select a Size
1 KG
$258.00
$258.00
Available to ship on28 April 2025Details
Select a Size
Change View
1 KG
$258.00
About This Item
UNSPSC Code:
42141501
NACRES:
NB.04
$258.00
Available to ship on28 April 2025Details
feature
degreases cotton wool
manufacturer/tradename
BRAND 28205
Related Categories
General description
Cotton plugs may prolong delivery time and may therefore influence the accuracy of measurement. In tests using pipettes that have had cotton plugs inserted by a cotton plugging machine, the results were within the indicated error limits.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)