CLS64061L
PYREX® economy Squibb separatory funnel
polyethylene stopper, PTFE stopcock, pack of 1 ea
Synonym(s):
pouring funnel
Select a Size
$294.00
Select a Size
About This Item
$294.00
Recommended Products
material
PTFE stopcock
polyethylene stopper
packaging
pack of 1 ea
manufacturer/tradename
Corning 6406-1L
body H × O.D.
395 mm × 119 mm
capacity
1 L
stem L × O.D.
52 mm × 7 mm
volume
1 L
1000 mL
Related Categories
General description
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)