CLS431843
Corning® centrifuge bottle
with standard screw cap, capacity 250 mL, polycarbonate
Synonym(s):
centrifugation bottles, digestion bottles, shaking bottles
Select a Size
$668.00
Select a Size
About This Item
$668.00
Recommended Products
material
clear bottle
flat bottom tube
polycarbonate
round bottle
wide-mouth bottle
sterility
non-sterile
feature
autoclavable
cap
packaging
pkg of 36 ea
manufacturer/tradename
Corning 431843
parameter
13500 × g max. RCF
capacity
250 mL
Related Categories
Features and Benefits
- Corning® centrifuge bottles have a specially designed cap for improved grip
- Includes clear graduations and textured marking spot.
- Bottles are designed with extra-thick walls for high durability
- Wide mouth for easier liquid filling
- Compatible with most centrifuges and do not require special buckets
- Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)