Skip to Content
Merck
All Photos(1)

Key Documents

CLS431134

Corning® Roller Bottles, CellBIND® Cell Culture Surface

clear polystyrene, sterile, bottle surface area 1700 cm2, cap (Easy Grip), case of 20

Synonym(s):

Corning roller bottles, roller bottles

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$589.00

$589.00


Please contact Customer Service [email protected] for availability.


Select a Size

Change View
20 EA
$589.00

About This Item

UNSPSC Code:
41122105
NACRES:
NB.13

$589.00


Please contact Customer Service [email protected] for availability.

material

cap (Easy Grip)
clear polystyrene
cylindrical bottle
high-density polyethylene cap
polystyrene bottle
wide-mouth bottle

description

expanded surface

sterility

sterile
sterile

feature

cap
disposable
graduations

packaging

case of 20

manufacturer/tradename

Corning 431134

bottle neck diam.

~45 mm

bottle surface area

1700 cm2

bottle working volume

340-510 mL

capacity

510 mL

diam. × H

117 mm × 271 mm

binding type

CellBIND® surface

Looking for similar products? Visit Product Comparison Guide

General description


  • The Corning CellBIND cell culture surface
  • Improves cell attachment leading to increased cell growth and yields
  • Results in more consistent and even cell attachment
  • Helps adapt cells to reduced serum or serum-free conditions
  • Reduces premature cell detachment from confluent cultures
Polystyrene roller bottles are also available with a Corning CellBIND surface treatment that uses a novel microwave plasma process for treating the culture surface. This process improves cell attachment by incorporating significantly more oxygen into the cell culture surface than the traditional plasma treatment, rendering it more hydrophilic and increasing the stability of the surface.

Features and Benefits

  • Easy Grip Caps feature large knurls designed for ergonomic handling
  • Easy Grip Vent Caps are designed for applications requiring consistent gas exchange
  • Bottles are manufactured from virgin polystyrene, caps from high density polyethylene
  • Both resins meet the USP Class VI requirements for plastic containers and closures
  • Treated with a Corning CellBIND surface treatment for enhanced cell attachment
  • One piece seamless construction with printed graduations
  • Printed lot numbers to aid in product traceability
  • Sterilized by g−radiation and certified non-pyrogenic

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service