Skip to Content
Merck
All Photos(1)

Key Documents

CLS4141

Corning® DeckWorks pipet tip station

0.1 - 10μL pipet tip station, extended length, graduated, non-sterile, polypropylene, 192 tips/layer, 5 layers/pack, 5 packs/case

Synonym(s):

pipet tip station, pipette tip station

Sign Into View Organizational & Contract Pricing

Select a Size

960 EA
$176.00
5 X 960 EA
$321.00

$176.00


Check Cart for Availability


Select a Size

Change View
960 EA
$176.00
5 X 960 EA
$321.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
volume:
0.1—10 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip shape: round
packaging:
pack of 960
manufacturer/tradename:
Corning 4141

$176.00


Check Cart for Availability

material

natural tip
polypropylene

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip shape: round

packaging

pack of 960

manufacturer/tradename

Corning 4141

volume range

0.1-10 μL

volume

0.1—10 μL

suitability

suitable for (pre- and post-PCR procedures, cell culture and assays requiring large tip quantities)

Looking for similar products? Visit Product Comparison Guide

General description

Corning® DeckWorks Pipet Tip Station

Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip stations contain 960 tips, are available in extended length 10 uL and traditional 200 uL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only.

Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities.

Corning DeckWorks tip stations are offered both sterile and nonsterile. This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.

The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.

Medical grade resins and production excellence allow for full certification as:

  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free

Legal Information

Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service