Skip to Content
Merck
All Photos(1)

Documents

CLS4149

Corning® DeckWorks low binding tips

1 - 300μL low binding pipet tips, graduated, hinged racks, natural, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case

Synonym(s):

low binding pipet tips, low binding pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

material

natural tip
polypropylene

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip shape: round

packaging

pack of 960

manufacturer/tradename

Corning 4149

volume range

1-300 μL

suitability

suitable for (quantitative assays in which sample binding can significantly alter results; specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.)

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® DeckWorks Low Binding Tips

Corning DeckWorks low binding tips are available in 10 uL, 200 uL and 300 uL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results.

Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides. This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.

The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.

Medical grade resins and production excellence allow for full certification as:

  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free

Legal Information

Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service