Skip to Content
Merck
All Photos(1)

Key Documents

CLS3965

Corning® 384 Well Deep Well Plate

clear polypropylene, round bottom, non-sterile, deep well, lid: no, pack of 5

Synonym(s):

384 well assay block, 384 well storage block

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$1,440.00

$1,440.00


Check Cart for Availability


Select a Size

Change View
100 EA
$1,440.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
clear bottom
clear polypropylene
polypropylene (plates, blocks and sealing mats)
round bottom
size:
384 wells

$1,440.00


Check Cart for Availability

material

clear bottom
clear polypropylene
polypropylene (plates, blocks and sealing mats)
round bottom

sterility

non-sterile

feature

lid: no
lid: no
skirt
deep well
plate format: 384 well storage block

packaging

case of 100
pack of 5

manufacturer/tradename

Corning 3965

size

384 wells

well volume

180 μL

color

clear

suitability

suitable for (general storage, liquid handling; automated applications)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • Certified DNase- and RNase-free
  • Total well volume 180 μL
Corning offers a complete range of polypropylene storage systems consisting of plates, blocks, and sealing mats. All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 384 well polypropylene plates and blocks are built to proposed industry standard footprint. All plates and blocks are automation-friendly and solvent resistant. The 384 well plates and blocks are certified DNase/RNase free and feature rounded bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service