Skip to Content
Merck
All Photos(1)

Key Documents

CLS3764BC

Corning® 384 well microplate, low flange

Tissue Culture (TC)-treated surface, black polystyrene, flat bottom, sterile, Generic Barcode, lid, pack of 20

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$663.00
100 EA
$3,220.00

$663.00


Check Cart for Availability


Select a Size

Change View
20 EA
$663.00
100 EA
$3,220.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
black polystyrene
clear bottom
flat bottom
polystyrene
polystyrene plate
size:
384 wells
384 wells

$663.00


Check Cart for Availability

material

black polystyrene
clear bottom
flat bottom
polystyrene
polystyrene plate

sterility

sterile

feature

lid
skirt
Generic Barcode
Low Flange

packaging

case of 100 plates
pack of 20

manufacturer/tradename

Corning 3764BC

maximum volume

112 μL

size

384 wells
384 wells

well volume

112 μL

well working volume

25-50 μL

working volume

80 μL

suitability

suitable for tissue

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 384 well microplate, low flange matrix active group TC-treated, black polystyrene has a flat bottom.
  • Sterile Black wall plates have low background fluorescence and minimal light scatter.
  • Opaque walls to prevent well-to-well crosstalk.
  • Direct microscopic viewing.
  • Sterilized by gamma radiation and nonpyrogenic.
  • Individual alphanumeric codes for good identification.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service