Skip to Content
Merck
All Photos(1)

Key Documents

CLS3729

Corning® 1536 well plates

flat bottom white polystyrene wells, NBS (TC-treated), 50/cs, non-sterile, lid: no

Synonym(s):

1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, HTS plate, TC-treated plate, immunoassay plate, tissue-culture treated plate

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$3,560.00

$3,560.00


Estimated to ship on21 May 2025



Select a Size

Change View
50 EA
$3,560.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom white polystyrene wells
polystyrene plate
white plate
size:
1536 wells

$3,560.00


Estimated to ship on21 May 2025


material

flat bottom white polystyrene wells
polystyrene plate
white plate

sterility

non-sterile

feature

lid: no
skirt
plate format: 1536 well standard

packaging

pack of 10
case of 50

manufacturer/tradename

Corning 3729

maximum volume

12.5 μL

size

1536 wells

working volume

8 μL

color

white

suitability

suitable for (fluorescent; luminescent assays)

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

Corning 1536 Well Plates

  • 1536 well polystyrene plates
  • Low background fluorescence, minimal light scatter and reduced crosstalk
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Opaque walls to prevent well-to-well crosstalk
  • Flat well bottoms possess superior flatness -minimizes variability within a plate and between plates
  • Round well bottoms for reduced air entrapment and improved CV′s and Z factor
  • Raised well bottoms for high sensitivity
  • Flood reservior on four sides to reduce instrument contamination
  • Suitable for fluorescent and luminescent assays
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers
  • NBS microplates are ideal for homogeneous assays with "sticky" enzymes or for reducing nonspecific binding to microplate wells
  • Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service