CLS351172
Corning® Falcon® 96 Well Cell Culture Plate
flat clear polystyrene, lid, sterile
Synonym(s):
96 Well Cell Culture Plate
Select a Size
$289.00
Select a Size
About This Item
$289.00
Recommended Products
material
flat clear polystyrene
sterility
sterile
feature
lid
manufacturer/tradename
Corning 351172
technique(s)
cell culture | mammalian: suitable
size
96 well
binding type
cell culture treated
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service