Skip to Content
Merck
All Photos(1)

Key Documents

CLS9018BC

Corning® 96 well plates

clear flat bottom, with generic barcode, polystyrene, high binding, without lid, sterile

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$1,480.00

$1,480.00


Available to ship on28 April 2025Details



Select a Size

Change View
100 EA
$1,480.00

About This Item

UNSPSC Code:
41121800
material:
clear bottom
flat bottom wells
size:
96 wells

$1,480.00


Available to ship on28 April 2025Details


material

clear bottom
flat bottom wells

sterility

non-sterile

feature

lid: no
skirt

packaging

pkg of 100 ea

manufacturer/tradename

Corning 9018BC

size

96 wells

well volume

360 μL

wells

96 wells

working volume

75-200 μL

suitability

suitable for (cell cuture)

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® is a leading brand in manufacturing best in class, high-quality, high-performance enzyme linked immunosorbent assay (ELISA) microplates for laboratory assays. These microplates have superior lot-to-lot and intra-well dimension consistency for reproducible and accurate results. ELISA microplates allow a common laboratory experiments to be performed on multiple samples at the same time.

Application

These plates are general assay plates, ideal for solution-based assays (i.e. hemagglutination or precipitation assays), serial dilutions and general storage applications.

Features and Benefits

  • Corning® 96-well ELISA microplate has high binding surface, which is ideal for immunoassays
  • High binding surface is capable to bind positively charged biomolecules (>10kD) with ionic groups and/or hydrophobic regions
  • Binding capacity of this surface is approximately 400 to 500ng IgG/cm²
  • 96-well plate format follows standard microplate footprint and dimensions for reliable results

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service