Key Documents
CLS3121STPK
Replacement stopcock, for Pyrex® small top desiccators with 24/29 standard taper stopcock
replacement 24/29 stopcock w/ hose connector, 1/cs
Synonym(s):
Corning stopcock
Select a Size
1 EA
$163.00
$163.00
Check Cart for Availability
Select a Size
Change View
1 EA
$163.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$163.00
Check Cart for Availability
material
borosilicate glass (borosilicate glass code 7740)
Joint
joint ST/NS24/29
feature
reusable
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 3121-STPK
H
117 mm
Related Categories
General description
TThis is a replacement 24/29 standard taper stopcock with a tooled #10 hose connection. The plug material is PTFE, and is a suitable replacement for CLS3121150, CLS3121200 and CLS3121250.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)