CLS3121200
Pyrex® large top desiccators with 24/29 standard taper stopcock
5.8L large top desiccator w/ 24/29 standard taper stopcock, 1/cs
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass (borosilicate glass code 7740)
feature
reusable
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 3121-200
H
296 mm
capacity
5.8 L
chamber D
115 mm
flange I.D.
223.5 mm
plate diam.
190 mm
Looking for similar products? Visit Product Comparison Guide
General description
This 5.8L PYREX desiccators has a rugged 24/29 standard taper stopcock with tooled hose connections that serve as the vacuum inlet. It is designed so that its manipulation is not affected by the vacuum. To introduce vacuum into the desiccator, loosen the stopcock to disengage the PTFE plug from contact with the glass. A half turn from this position closes the desiccator. This design eliminates the need for grease. The tubulation O.D. is approximately 10mm. Approximate bowl volume capacity is 5800mL. Warning: Do not heat or subject to pressure. Rated 1- atmosphere vacuum.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service