CLS3081150LO
Pyrex® small knob top desiccator
Lower only
Synonym(s):
equipment replacement accessory
Select a Size
$329.00
Estimated to ship on16 May 2025
Select a Size
About This Item
$329.00
Estimated to ship on16 May 2025
Recommended Products
material
borosilicate glass
manufacturer/tradename
Corning 3081-150LO
Related Categories
General description
Warning: Do not heat or subject to pressure. Not for vacuum applications.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)