Key Documents
CLS285522
Corning® cover glasses
square, No. 2, W × L 22 mm × 22 mm
Synonym(s):
coverslips, glass covers, slide covers
Select a Size
100 EA
$30.20
1000 EA
$171.00
$30.20
Available to ship on22 April 2025Details
Select a Size
Change View
100 EA
$30.20
1000 EA
$171.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.16
$30.20
Available to ship on22 April 2025Details
material
borosilicate glass
square
packaging
pack of 100 ea
case of 1000 ea
manufacturer/tradename
Corning 2855-22
W × L
22 mm × 22 mm
binding type
non-treated surface
Features and Benefits
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1