CLS285018
Corning® cover glasses
square, No. 11/2, W × L 18 mm × 18 mm
Synonym(s):
coverslips, glass covers, slide covers
Select a Size
$40.80
Available to ship on14 April 2025Details
Select a Size
About This Item
$40.80
Available to ship on14 April 2025Details
Recommended Products
material
borosilicate glass
square
packaging
pack of 200 ea
case of 2000 ea
manufacturer/tradename
Corning 2850-18
W × L
18 mm × 18 mm
binding type
non-treated surface
Features and Benefits
- Corning® cover glasses are made from optically clear borosilicate glass.
- Chemically resistant cover glasses are not affected by weathering or surface attachment.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)