Established from a bone marrow biopsy of a 22-month-old male with disseminated neuroblastoma in 1972. The morphology varies with sub-population from cells with long processes through small, flattened, epithelioid cells. Dense cultures tend to form loosely adherent aggregates. They are multipotential with regard to neuronal enzyme expression and display a high capacity to convert tyrosine to dopamine.
Application
Chemotherapy development, chromosome studies, drug resistance, differentiation studies.
Split sub-confluent cultures (70-80%) 1:10 to 1:25 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)