Skip to Content
Merck
All Photos(1)

Key Documents

SK-N-AS

94092302, human nerve (bone marrow metastasis), Polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

SK-N-AS, 94092302

biological source

human nerve (bone marrow metastasis)

growth mode

Adherent

karyotype

46

morphology

Polygonal

products

Not specified

receptors

Vimentin

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human neuroblastoma

Cell Line Description

Derived from a female patient with neuroblastoma from the metastasis of the bone marrow. Tumourigenic in nude mice.

Application

Neuroblastoma differentiation studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,12
D13S317: 9
D16S539: 14
D5S818: 11,12
D7S820: 11,13
THO1: 9.3
TPOX: 11,12
vWA: 16,17

Culture Medium

DMEM or EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service