Skip to Content
Merck
All Photos(1)

Documents

RK-13 Cell Line from rabbit

21715, rabbit kidney, Epithelial-like

Synonym(s):

GL RK/13 Cells, Glaxo Laboratories Rabbit Kidney 13 Cells, R13 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

RK-13 Cell Line from rabbit, rabbit kidney, 00021715

biological source

rabbit kidney

growth mode

Adherent

karyotype

2n=44, stem-line = 56 (=22%)

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rabbit kidney

Cell Line Description

RK13 has been derived from kidney cells of a 5 week old rabbit. The cell line has been shown to be susceptible to pseudorabies virus, rabbitpox virus, myxomatose virus, vaccinia and EAV. Sensitivity to other viruses such as Simian adenovirus and rubella has been reported as well with the production of cytopathic effect (CPE). RK13 has been successfully treated with thiamutin for eradication of mycoplasma. The Bovine Viral Diarrhoea Virus (BVDV) status of this cell line was previously described as negative. Results of recent multiple molecular tests for BVDV that were undertaken by two independent testing laboratories were equivocal. This means that we cannot confirm the BVDV status of this cell line.

Application

Virus studies

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37ºC; monolayer is confluent after 48-56 hrs and stable for at least 21d; small syncytia in elder cultures are typical.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service