Skip to Content
Merck
All Photos(1)

Key Documents

C8742

Sigma-Aldrich

CHIC-35

≥97% (chiral, HPLC)

Synonym(s):

(6S)-2-Chloro-5,6,7,8,9,10-hexahydro-cyclohept[b]indole-6-carboxamide, Compound (S)-35

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$175.00
5 MG
$688.00

$175.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
1 MG
$175.00
5 MG
$688.00

About This Item

Empirical Formula (Hill Notation):
C14H15ClN2O
CAS Number:
Molecular Weight:
262.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$175.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥97% (chiral, HPLC)

form

solid

solubility

DMSO: >10 mg/mL
H2O: <2 mg/mL

storage temp.

2-8°C

SMILES string

ClC1=CC=C(NC2=C3CCCC[C@@H]2C(N)=O)C3=C1

InChI

1S/C14H15ClN2O/c15-8-5-6-12-11(7-8)9-3-1-2-4-10(14(16)18)13(9)17-12/h5-7,10,17H,1-4H2,(H2,16,18)/t10-/m0/s1

InChI key

ABIVOOWWGYJNLV-JTQLQIEISA-N

Biochem/physiol Actions

CHIC-35 is cell-permeable, metabolically stable, and very potent inhibitor of SIRT1.
CHIC-35 is cell-permeable, metabolically stable, and very potent inhibitor of SIRT1; IC50 of S-isomer is 60 nM; IC50 of mixed isomers is 124 nM. There is no inhibition of SIRT3 or HDAC. The IC50 for SIRT2 is 2.77 μM. Sirtuins are protein deacetylases, which represent a new class of histone deacetylases (HDAC) involved in gene silencing. SIRT modulators are potential therapeutics for cancer, diabetes, muscle differentiation, heart failure, neurodegeneration, and aging.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service