Skip to Content
Merck
All Photos(1)

Key Documents

C5612

Sigma-Aldrich

Anti-CD30 Ligand antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CD30L

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

indirect ELISA: 0.5-1.0 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Tnfsf8(21949)

General description

CD30 Ligand (CD30L) is a type II membrane glycoprotein of the tumor necrosis factor (TNF) superfamily. It is expressed on the cell surface of activated T cells, B cells, and monocytes. CD30L is also present in other cells, including mature dendritic cells, macrophages and mast cells. CD30L is also reffered to as Tnfsf8 (Tumor necrosis factor ligand superfamily member 8).

Immunogen

purified recombinant mouse CD30 ligand (amino acids 68-239) extracellular domain, expressed in NSO cells.

Application

Anti-CD30 antibody produced in goat is suitable for indirect ELISA at a concentration of 0.5-1.0μg/mL and western blotting at a concentration of 0.1-0.2μg/mL.

Biochem/physiol Actions

CD30L (CD30 ligand) along with CD30 is involved in activation and proliferation of T and B cells. They are also involved in the regulation of initial artherosclerosis development. CD30L/CD30 is crucial for maintenance and activation of IL (Interleukin)-17A-producing γδ T cells. CD30L/CD30 causes down-regulation of IL-2, resulting in Th17 differentiation of CD4+ T cells. CD30L helps in memory phenotype CD4+ T cells-mediated protection against infection with Listeria monocytogenes. CD30L/CD30 signaling is critical for Th1 response against Mycobacterium bovis bacillus Calmette-Guérin (BCG) infection. They are also linked to allergic rhinitis pathogenesis in mice. CD30L/CD30 signaling is involved in pathogenesis of inflammatory bowel diseases.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Sun et al.
Mucosal immunology, 6(6), 1191-1201 (2013-04-04)
CD30 ligand (CD30L, CD153), a member of the tumor necrosis factor (TNF) superfamily, and its receptor CD30 are important for differentiation and activation of CD4(+) T helper type 17 (Th17) cells. In this report, we demonstrate that the interleukin 17A
Takafumi Fuchiwaki et al.
European journal of immunology, 41(10), 2947-2954 (2011-07-09)
CD30 ligand (CD30L) plays an important role in the amplification and/or activation of effector CD4(+) T cells, irrespective of Th cell subset. To examine the role of CD30L in allergic rhinitis, we evaluated an OVA model of allergic rhinitis in
Ce Tang et al.
Journal of immunology (Baltimore, Md. : 1950), 181(9), 6316-6327 (2008-10-23)
A CD30 ligand (CD30L, CD153) is a type II membrane-associated glycoprotein belonging to the TNF family. To illustrate the potential role of CD30L in CD4(+) Th1 cell responses, we investigated the fate of Ag-specific CD4(+) T cells in CD30L-deficient (CD30L(-/-))
H J Gruss et al.
The American journal of pathology, 149(2), 469-481 (1996-08-01)
The CD30 ligand (CD30L) is a type II transmembrane glycoprotein of the tumor necrosis factor ligand superfamily. Recent cloning of CD30L has enabled studies to explore its function and tissue distribution. For instance, recombinant CD30L has been shown to co-stimulate
Amanda C Foks et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(12), 2862-2868 (2012-10-23)
Costimulatory molecules tightly control immune responses by providing positive signals that promote T-cell activation or by transducing inhibitory signals that limit T-cell responses. CD30 and CD30L are members of the tumor necrosis factor receptor superfamily and are involved in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service