- The central role of CD30L/CD30 interactions in allergic rhinitis pathogenesis in mice.
The central role of CD30L/CD30 interactions in allergic rhinitis pathogenesis in mice.
CD30 ligand (CD30L) plays an important role in the amplification and/or activation of effector CD4(+) T cells, irrespective of Th cell subset. To examine the role of CD30L in allergic rhinitis, we evaluated an OVA model of allergic rhinitis in CD30L knock out (KO) mice on a BALB/c background sensitized with OVA. Symptoms of allergic rhinitis such as eosinophil infiltration into the nasal mucosa were drastically diminished in OVA-sensitized CD30L KO mice following intranasal challenge with OVA. The levels of OVA-specific IgE in the sera and the Th2 response in nasopharynx-associated lymphoid tissues and cervical LNs of CD30L KO mice were significantly lower than those of WT mice following intranasal challenge with OVA. Intranasal administration of CD30-Ig during the effector phase with OVA significantly prevented the development of allergic rhinitis in WT mice. These results suggest that CD30L plays an important role in allergic rhinitis and that the inhibition of CD30L/CD30 signaling might be useful as a novel biological therapy for allergic rhinitis.