Skip to Content
Merck
All Photos(1)

Key Documents

C1049

Sigma-Aldrich

(+)-Calcium L-tartrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4O6Ca · xH2O
CAS Number:
Molecular Weight:
188.15 (anhydrous basis)
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

O[C@@H]1[C@@H](O)C(=O)O[Ca]OC1=O

InChI

1S/C4H6O6.Ca/c5-1(3(7)8)2(6)4(9)10;/h1-2,5-6H,(H,7,8)(H,9,10);/q;+2/p-2/t1-,2-;/m1./s1

InChI key

GUPPESBEIQALOS-ZVGUSBNCSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel E Rusyniak et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 9(1), 79-81 (2012-08-29)
Cream of tartar (potassium bitartrate) has a long history as a cooking aid and medicinal purgative. Despite containing large amounts of potassium, there are no well-documented cases of it causing toxicity. We report two cases in which intentional ingestions of
Anne Toboldt et al.
Applied and environmental microbiology, 78(20), 7347-7357 (2012-08-14)
In this study, the population structure, incidence, and potential sources of human infection caused by the d-tartrate-fermenting variant of Salmonella enterica serovar Paratyphi B [S. Paratyphi B (dT+)] was investigated. In Germany, the serovar is frequently isolated from broilers. Therefore
Gergely Varga et al.
Bioorganic & medicinal chemistry letters, 23(6), 1789-1792 (2013-02-12)
Di-O-cinnamoylated, -p-coumaroylated, and -feruloylated d-, l- and meso-tartaric acids were synthesized as analogues of the natural product FR258900, a glycogen phosphorylase (GP) inhibitor with in vivo antihyperglycaemic activity. The new compounds inhibited rabbit muscle GP in the low micromolar range
Andrea Bencini et al.
Chemical communications (Cambridge, England), 48(84), 10428-10430 (2012-09-18)
A chiral ditopic polyammonium receptor featuring two [9]aneN(3) moieties separated by a (S)-BINOL linker is able to selectively bind and sense in water (S,S)-tartaric acid over its (R,R)/meso forms.
J W H Smith et al.
Journal of hazardous materials, 235-236, 279-285 (2012-09-07)
Impregnated activated carbons (IACs) that are used in multi-gas respirator applications usually contain copper and/or zinc impregnants. Co-impregnating with properly selected acids can improve the distribution of the metallic impregnant on the carbon and improve the gas adsorption capacity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service