Skip to Content
Merck
All Photos(1)

Key Documents

B8061

Sigma-Aldrich

N-Butyl-N-(4-hydroxybutyl)nitrosamine

ISOPAC®, ≥90% (GC)

Synonym(s):

BBN, N-Butyl-N-butan-4-ol-nitrosamine, N-Butyl-N-nitroso-4-aminobutanol, OH-BBN

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$111.00

$111.00


Estimated to ship on24 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
$111.00

About This Item

Empirical Formula (Hill Notation):
C8H18N2O2
CAS Number:
Molecular Weight:
174.24
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.25

$111.00


Estimated to ship on24 March 2025


Request a Bulk Order

Quality Level

Assay

≥90% (GC)

form

liquid

storage temp.

2-8°C

SMILES string

N(N=O)(CCCCO)CCCC

InChI

1S/C8H18N2O2/c1-2-3-6-10(9-12)7-4-5-8-11/h11H,2-8H2,1H3

InChI key

DIKPQFXYECAYPC-UHFFFAOYSA-N

Application

N-Butyl-N-(4-hydroxybutyl)nitrosamine (BBN), a carcinogen, is used to induce histiologically relevant aggressive urinary bladder cancer in animal models.

Biochem/physiol Actions

Carcinogen used to induce urinary bladder cancer in animal models. The result of exposure is histologically comparable to human urinary bladder tumorigenesis. Used in chemopreventative studies.

Packaging

Packaged in a 100 mL serum bottle with butyl rubber stopper and aluminum tear seal.

Caution

Injecting any compatible solvent permits preparation of any desired strength solution without exposure.

Reconstitution

Dissolving the contents in 100 mL of solvent yields a 1% solution.

Legal Information

Isopac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

296.6 °F

Flash Point(C)

147 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

João C Fernandes et al.
Marine drugs, 10(12), 2661-2675 (2013-01-25)
Urinary bladder cancer is one of the most common cancers worldwide, with the highest incidence in industrialized countries. Patients with cancer commonly use unconventional and complementary therapy including nutraceuticals. In this study we evaluated the efficacy of chitooligosaccharides (in orange
Yuji Sagara et al.
Cancer epidemiology, 34(3), 350-354 (2010-04-07)
Green tea polyphenol (GTP) suppresses malignancy in bladder cancer cell lines. However, the detail of its anti-carcinogenic effect in vivo is not fully understood. This study investigated the effect of GTP on bladder tumor size and angiogenesis in mice given
Koji Nishizawa et al.
International journal of cancer, 127(5), 1180-1187 (2009-12-30)
We previously reported that the expression of CXC chemokine receptor-4 (CXCR4) was upregulated in invasive bladder cancers and that the small peptide T140 was a highly sensitive antagonist for CXCR4. In this study, we identified that CXCR4 expression was induced
C Vasconcelos-Nóbrega et al.
In vivo (Athens, Greece), 26(4), 727-739 (2012-07-10)
Experimental urinary bladder tumours have been proposed as a useful model for the study of urinary bladder carcinogenesis, as well as for evaluating new therapeutic strategies. Consequently, the administration of chemical carcinogens is one of the most commonly used methods
Xiao-Li Xie et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(11), 3934-3940 (2012-08-15)
In the present study, effects of L-leucine and L-isoleucine on rat bladder carcinogenesis were investigated using AIN-93G and MF basal diet. In Experiment 1, N-butyl-N-(4-hydroxybutyl)-nitrosamine was used as an initiator of bladder carcinogenesis. In the AIN-93G diet groups, a significantly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service