Skip to Content
Merck
All Photos(1)

Documents

B7024

Sigma-Aldrich

Anti-Goat IgG (whole molecule)–Biotin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

goat

technique(s)

direct ELISA: 1:20,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The antibody binds to all goat Igs.

Application

Anti-Goat IgG (whole molecule)-Biotin antibody produced in rabbit has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunohistochemistry
  • immunofluorescence analysis

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of biotin to Anti-Goat IgG (whole molecule) antibody allows for the binding of various labels such as avidin or streptavidin.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Preparation Note

Adsorbed to reduce background staining with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regional and cellular distribution of ephrin-B1 in adult mouse brain
Migani P, et al.
Brain Research, 1247(4), 50-61 (2009)
J Martin Wild
The Journal of comparative neurology, 525(7), 1731-1742 (2017-01-13)
The dorsal pallidum in birds is considered similar, if not homologous, to the globus pallidus (GP) of mammals. The dorsal pallidum projects to both thalamic and midbrain targets similar to the direct and indirect pathways arising from the internal and
Immunoglobulin G, complement factor C3 and lymphocytes in proliferative intraocular disorders
Weller M, et al.
International Ophthalmology, 14(4), 277-283 (1990)
A real-time immuno-PCR assay for routine ultrasensitive quantification of proteins
Adler M, et al.
Biochemical and Biophysical Research Communications, 308(2), 240-250 (2003)
Elina V García et al.
Theriogenology, 81(8), 1032-1041 (2014-03-04)
Bone morphogenetic proteins (BMPs) play a crucial role in mammalian reproduction, but little is known about their expression and function in the oviduct, where preimplantation events take place. In the present study, messenger RNA (mRNA) expression of BMPs was examined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service