AXYVT38410ULR
Pipette tips in 384-tip format, robotic
size 10 μL, pkg of 5x3840tips/cs, racked
Synonym(s):
Automated pipette tips, Robotic pipette tips
About This Item
RNase free
barrier: no
endotoxin free
filter: no
Corning
Recommended Products
material
clear
sterility
non-sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
manufacturer/tradename
Axygen Inc VT-384-10UL-R
Corning
packaging
pkg of 5x3840tips/cs
size
10 μL
volume
10 μL
Looking for similar products? Visit Product Comparison Guide
General description
- Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
- Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
- The clear polypropylene high-quality tips are ideal for sensitive biological applications.
- RNase-/DNase-free and nonpyrogenic.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service