Z286400
FEP sample tube liner
Wilmad 6005-8, O.D. 5 mm
Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes
All Photos(1)
About This Item
Recommended Products
description
Recommended outer tube 528-PP-8
packaging
pack of 1 ea
manufacturer/tradename
Wilmad 6005-8
O.D.
5 mm
volume
80 μL
General description
Thin-wall FEP liner minimizes filling factor losses of using inserts and protects NMR tube from etching effects of strong acids containing fluoride ions. Liners are 8 in. L and fit snugly into most common 7 in. L thin-wall tubes. A PTFE plug to close each liner is included. Not recommended for elevated temperature studies.