Skip to Content
Merck
All Photos(1)

Key Documents

AXYTF205WBRS

Pipette tips

capacity 200 μL, wide bore, filtered, clear, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
200 μL
sterility:
γ-irradiated
sterile
feature:
DNA free
RNase free
barrier
endotoxin free
filter
wide bore
packaging:
case of 960 tips racked
manufacturer/tradename:
Axygen Inc
Corning TF-205-WB-R-S
Product AXYTF205WBRS is not currently sold in your country. Contact Technical Service

material

clear

sterility

γ-irradiated
sterile

feature

DNA free
RNase free
barrier
endotoxin free
filter
wide bore

packaging

case of 960 tips racked

manufacturer/tradename

Axygen Inc
Corning TF-205-WB-R-S

capacity

200 μL

volume

200 μL

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service