Skip to Content
Merck
All Photos(1)

Key Documents

AXYST200

Screw cap tubes without caps, conical bottom

size 2.0 mL, clear, pkg of 8x500caps/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Product AXYST200 is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Axygen Inc ST-200
Corning

packaging

pkg of 8x500caps/cs

size

2.0 mL

color

clear

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service