Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR0208FCPC

PCR strip tubes

with flat strip caps with writing surface, thin wall, 8 per strip, size 0.2 mL, clear, pkg of 10x125strips/cs

Synonym(s):

96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Product AXYPCR0208FCPC is not currently sold in your country. Contact Technical Service

material

cap (flat strip caps)
polypropylene

sterility

non-sterile

manufacturer/tradename

Axygen Inc PCR-0208-FCP-C
Corning

packaging

pkg of 10x125strips/cs

size

0.2 mL

color

clear
clear

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® PCR strip tubes with flat strip caps are designed to fit a wide variety of thermal cyclers.
  • Caps provide a leak-proof seal for sample protection.
  • Uniform ultra-thin wall thickness facilitates optimal thermal transfer.
  • Flat caps feature a clear surface ideal for fluorescent luminescent and real-time PCR applications.

Legal Information

Axygen is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service