Skip to Content
Merck
All Photos(2)

Key Documents

AXYHGB1530MC15

Corning® Axygen® 30 well MC comb for use with 15 cm gel system

clear, thickness (1.5 mm), non-sterile

Synonym(s):

gel system accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.15
Product AXYHGB1530MC15 is not currently sold in your country. Contact Technical Service

sterility

non-sterile

manufacturer/tradename

Corning HGB15-30MC-15

thickness

(1.5 mm)

color

clear

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service