Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500989

Sigma-Aldrich

Anti-FOLR1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FR-α, Folate receptor, Folate receptor α, Folate receptor 1, adult

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$693.00

$693.00


Check Cart for Availability


Select a Size

Change View
100 μG
$693.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$693.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 29 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOLR1(2348)

General description

The folate receptor 1 (FOLR1) gene encodes for folate receptor α protein, a membrane-bound glycoprotein. The FOLR1 gene is mapped on the human chromosome at 11q13.4. Anti-FOLR1 antibody detects endogenous levels of total FOLR1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human FOLR1.

Immunogen Range: 41-90

Biochem/physiol Actions

Folate receptor α protein binds and facilitates the transport of folate into the cells. This protein acts as a therapeutic target as well as a prognostic marker for various kinds of cancers. Upregulation of the FOLR1 gene is associated with ovarian, lung, breast, brain, colorectal, and gastric cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minsung Kim et al.
PloS one, 13(6), e0198347-e0198347 (2018-06-07)
Gastric cancer is a malignancy that has a high mortality rate. Although progress has been made in the treatment of gastric cancer, many patients experience cancer recurrence and metastasis. Folate receptor 1 (FOLR1) is overexpressed on the cell surface in
Michelle R O'Byrne et al.
Birth defects research. Part A, Clinical and molecular teratology, 88(8), 689-694 (2010-08-05)
Meningomyelocele (MM) results from lack of closure of the neural tube during embryologic development. Periconceptional folic acid supplementation is a modifier of MM risk in humans, leading toan interest in the folate transport genes as potential candidates for association to
Yuan Yuan et al.
Human pathology, 40(10), 1453-1460 (2009-05-21)
The objective of this study was to analyze the diagnostic and clinical role of the folate receptor-alpha (FOLR1) and folate receptor-gamma (FOLR3) genes in effusion cytology. Expression of the FOLR1 protein product, FR-alpha, was additionally studied. Ninety-one effusions (71 ovarian
Correction: Folate receptor-α (FOLR1) expression and function in triple negative tumors.
Brian M Necela et al.
PloS one, 10(4), e0127133-e0127133 (2015-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service