Skip to Content
Merck
All Photos(1)

Key Documents

AV49937

Sigma-Aldrich

Anti-ADAM33 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ADAM metallopeptidase domain 33, Anti-DJ964F7.1, Anti-DKFZp434K0521, Anti-FLJ35308, Anti-FLJ36751, Anti-MGC149823, Anti-MGC71889

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62 kDa

species reactivity

dog, rat, human, mouse, pig, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM33(80332)

General description

ADAM metallopeptidase domain 33 (ADAM33) is a member of the ADAM (a disintegrin and metalloprotease domain) family. ADAM33 is largely expressed in mesenchymal cells including airway fibroblasts, myofibroblasts, and smooth muscle cells.

Immunogen

Synthetic peptide directed towards the middle region of human ADAM33

Application

Anti-ADAM33 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/mL.

Biochem/physiol Actions

The members of the ADAM (a disintegrin and metalloprotease domain) family are involved in cell-to-cell and cell-to-matrix interactions, neurogenesis and muscle development. The expression of ADAM33 has been linked to asthma and chronic obstructive pulmonary disease (characterised by chronic bronchitis and emphysema). ADAM33 is also associated with inflammation of the lungs which is activated against etiological viral agents.

Sequence

Synthetic peptide located within the following region: HDSAQLLTGRAFQGATVGLAPVEGMCRAESSGGVSTDHSELPIGAAATMA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deng-Chuan Zhou et al.
Molecular biology reports, 42(2), 409-422 (2014-10-05)
A series of observational studies have been made to investigate the association of the ADAM33 gene polymorphisms with the risk of COPD, but their results were conflicting. Therefore, we performed an updated meta-analysis to quantitatively summarize the associations of ADAM33
Shelby P Umland et al.
American journal of respiratory cell and molecular biology, 29(5), 571-582 (2003-06-05)
We examined transcript expression and post-transcriptional regulation of human ADAM33, a recently identified asthma gene. A detailed messenger RNA (mRNA) expression profile was obtained using Northern, reverse transcription polymerase chain reaction, and in situ hybridization analyses. ADAM33 mRNA was expressed
Emanuele Baurakiades et al.
Journal of clinical virology : the official publication of the Pan American Society for Clinical Virology, 61(4), 585-589 (2014-12-03)
ADAM28, ADAM33, IL-13, IL-4 and other cytokines (IL-6 and IL-10) seem to play important roles in the persistence and maintenance of acute inflammatory processes that ultimately lead to lung remodeling and pulmonary fibrosis, which may be responsible for the high
Feng Lin et al.
Molecular medicine reports, 8(4), 1209-1215 (2013-08-13)
A disintegrin and metalloproteinase 33 (ADAM33) has been identified as an asthma susceptibility gene; however, the role of ADAM33 in the pathogenesis and progression of asthma remains to be elucidated. As ADAM33 is predominantly expressed in airway smooth muscle cells
Paul Van Eerdewegh et al.
Nature, 418(6896), 426-430 (2002-07-12)
Asthma is a common respiratory disorder characterized by recurrent episodes of coughing, wheezing and breathlessness. Although environmental factors such as allergen exposure are risk factors in the development of asthma, both twin and family studies point to a strong genetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service