Skip to Content
Merck
All Photos(1)

Key Documents

AV45043

Sigma-Aldrich

Anti-ACVR2B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Acvr2B Antibody, Acvr2B Antibody - Anti-ACVR2B antibody produced in rabbit, Anti-Activin A receptor, type IIB

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$609.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

dog, guinea pig, human, horse, sheep, bovine, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACVR2B(93)

Immunogen

Synthetic peptide directed towards the C terminal region of human ACVR2B

Application

Anti-ACVR2B antibody produced in rabbit is suitable for western blotting at a concentration of 0.25μg/ml.

Biochem/physiol Actions

Activin A receptor, type IIB (ACVR2B) binds activin ligands that promote growth and differentiation via the TGF-β signaling. ACVR2B is expressed in adult hypothalamus and forebrain. Activin receptors IIA and IIB may regulate germ cell proliferation in the human ovary during primordial follicle formation and in mouse testis.

Sequence

Synthetic peptide located within the following region: HDAEARLSAGCVEERVSLIRRSVNGTTSDCLVSLVTSVTNVDLPPKESSI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Miller et al.
Journal of neuroendocrinology, 24(6), 962-972 (2012-02-03)
The inhibin/activin family of proteins is known to have a broad distribution of synthesis and expression in many species, as well as a variety of functions in reproductive and other physiological systems. Yet, our knowledge regarding the production and function
Sirisha H S Mendis et al.
Biology of reproduction, 84(2), 379-391 (2010-10-12)
Activin affects many aspects of cellular development, including those essential for reproductive fitness. This study examined the contribution of activin A to murine fetal testicular development, revealing contrasting outcomes of activin actions on Sertoli cells and gonocytes. Shortly after sex
S J Martins da Silva et al.
Developmental biology, 266(2), 334-345 (2004-01-24)
The formation of the essential functional unit of the ovary, the primordial follicle, occurs during fetal life in humans. Factors regulating oogonial proliferation and interaction with somatic cells before primordial follicle formation are largely unknown. We have investigated the expression
Se-Jin Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(50), 18117-18122 (2005-12-07)
Myostatin is a secreted protein that normally functions as a negative regulator of muscle growth. Agents capable of blocking the myostatin signaling pathway could have important applications for treating human muscle degenerative diseases as well as for enhancing livestock production.
Varuna C Banduseela et al.
Physiological genomics, 45(12), 477-486 (2013-04-11)
Critical illness myopathy (CIM) is characterized by a preferential loss of the motor protein myosin, muscle wasting, and impaired muscle function in critically ill intensive care unit (ICU) patients. CIM is associated with severe morbidity and mortality and has a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service