Skip to Content
Merck
All Photos(1)

Key Documents

AV48246

Sigma-Aldrich

Anti-RAB11B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-H-YPT3, Anti-MGC133246, Anti-RAB11B, member RAS oncogene family

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

24 kDa

species reactivity

bovine, rat, mouse, goat, dog, human, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB11B(9230)

General description

RAB11B is a GTP-binding protein that regulates diverse cellular functions.It is expressed in vesicular compartments in parietal and epithelial cells. Neuronal Rab11b has been implicated in exocytosis.
Rabbit Anti-RAB11B antibody recognizes zebrafish, human, mouse, rat, canine, chicken, and bovine RAB11B.

Immunogen

Synthetic peptide directed towards the C terminal region of human RAB11B

Application

Rabbit Anti-RAB11B antibody is suitable for western blot applications at a concentration of 0.25 μg/ml.

Biochem/physiol Actions

RAB11B possesses GTPase activity.

Sequence

Synthetic peptide located within the following region: IETSALDSTNVEEAFKNILTEIYRIVSQKQIADRAAHDESPGNNVVDISV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lynne A Lapierre et al.
Experimental cell research, 290(2), 322-331 (2003-10-22)
The Rab11 family of small GTPases is composed of three members, Rab11a, Rab11b, and Rab25. While recent work on Rab11a and Rab25 has yielded some insights into their function, Rab11b has received little attention. Therefore, we sought to examine the
Mikhail V Khvotchev et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 23(33), 10531-10539 (2003-11-25)
Using PC12 cells that express transfected human growth hormone (hGH) as a secreted reporter protein, we have searched for Rab proteins that function in exocytosis. Among the Rab proteins tested, we found that besides the previously described Rab3 proteins, only

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service