Skip to Content
Merck
All Photos(12)

Key Documents

ZRB2443

Sigma-Aldrich

Anti-PDGF-C Antibody, clone 4D14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$357.00
4 X 25 μL
$735.00

$357.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$357.00
4 X 25 μL
$735.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$357.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D14, recombinant monoclonal

description

4D14 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 12.5 kDa
observed mol wt ~25 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PDGFC(56034)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4D14 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Platelet-derived growth factor C (PDGF-C). It targets an epitope within 20 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal half of human Platelet-derived growth factor C.

Application

Quality Control Testing

Evaluated by Western Blotting in PDGF-C recombinant protein fragment.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected human recombinant fragment of PDGF-C expressed in E. coli.

Tested applications

Immunocytochemistry Analysis: A 1:300 dilution from a representative lot detected PDGF-C in A549 cells.

Affinity Binding Assay: A representative lot of this antibody bound PDGF-C with a KD of 4.8 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PDGF-C in human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Platelet-derived growth factor C (UniProt: Q9NRA1; also known as PDGF-C, Fallotein, Spinal cord-derived growth factor, SCDGF, VEGF-E) is encoded by the PDGFC (also known as SCDGE, UNQ174/PRO200) gene (Gene ID: 56034) in human. PDGF-C is a disulfide-linked homodimeric growth factor that is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off to produce the mature form. It plays an essential role in the regulation of embryonic development, cell proliferation, cell migration, survival and chemotaxis. It also plays an important role in wound healing and angiogenesis. It serves as a potent mitogen and chemoattractant for cells of mesenchymal origin. Its CUB domain is localized to amino acids 46 to 163. Proteolytic removal of the N-terminal CUB domain is essential for unmasking the receptor-binding epitopes of the core dom+D8ain. Cleavage after basic residues in the hinge region (region connecting the CUB and growth factor domains) gives rise to the receptor-binding form. PDGF-C expression is observed in the fallopian tube, vascular smooth muscle cells in kidney, breast and colon tissue, and in visceral smooth muscle of the gastrointestinal tract. It is highly expressed in retinal pigment epithelia. In the kidney, it is constitutively expressed in parietal epithelial cells of Bowman′s capsule, tubular epithelial cells, and in arterial endothelial cells. Higher levels of PDGF-C and have been reported in the malignant breast tumor microenvironment. In MCF-7 cells increased expression of PDGF-C is reported to enhance cell proliferation and tumor cell motility. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hurst, NJ., et al. (2012). Biochem. J. 441(3); 909-918; Gilbertson, DJ., et al. (2001). J. Biol. Chem. 276(29); 27406-27414).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service