Skip to Content
Merck
All Photos(3)

Key Documents

SAB5700596

Sigma-Aldrich

Recombinant 2019-nCoV Nucleocapsid Protein with His tag

Synonym(s):

Nucleocapsid protein, Nucleoprotein, Protein N

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$808.00

$808.00


Check Cart for Availability


Select a Size

Change View
100 μG
$808.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.26

$808.00


Check Cart for Availability

recombinant

expressed in E. coli

Quality Level

Assay

>95% (HPLC)
>95% (SDS-PAGE)

form

liquid

mol wt

49.4 kDa

impurities

<1.0 EU/μg protein (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

SARS virus ... N(43740575)

General description

Recombinant SARS-COV-2(2019-nCoV) Nucleocapsid Protein is produced by E. coli expression system. The target protein is expressed with sequence (Met1-Ala419) of SARS-COV-2(2019-nCoV) Nucleocapsid (Accession # QHD43423.2) fused with an 6XHis tag at the N-terminus.

Biochem/physiol Actions

Measured by its binding ability in a functional ELISA. Immobilized Recombinant SARS-COV-2 Nucleocapsid at 2μg/mL (100 μL/well) can bind Anti-Nucleocapsid antibody with a linear range of 0.12-1.64 ng/mL. Measured by its binding ability in a functional ELISA. Immobilized Recombinant SARS-CoV-2 envelope Protein at 2μg/mL (100 μL/well) can bind Recombinant SARS-CoV-2 Nucleocapsid protein, the EC50 of Recombinant SARS-CoV-2 Nucleocapsid protein is 41.10 ng/mL.

Reconstitution

For lyophilized protein : Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water.

Storage and Stability

Store the lyophilized protein at -20°C to -80°C for long term. After reconstitution, the protein solution is stable at -20°C for 3 months, at 2-8 °C for up to 1 week. This product is stable at ≤ -70°C for up to 6 months from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Data presented is the available current product information and provided as-is. These products have not been tested or verified in any additional applications, sample types, including any clinical use. Experimental conditions must be empirically derived by the user. Our Antibody Guarantee only covers tested applications stated herein and conditions presented in our product information and is not extended to publications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service