Skip to Content
Merck
All Photos(1)

Key Documents

AV42287

Sigma-Aldrich

Anti-FST antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FS, Anti-Follistatin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Check Cart for Availability


Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

sheep, goat, rat, dog, mouse, human, horse, rabbit, guinea pig, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FST(10468)

General description

Follistatin is an autocrine glycoprotein component of the inhibin-activin-follistatin axis that mediates many cellular processes via its ability to bind and neutralize members of the TGF-β superfamily such as activin and myostatin/GDF-8.

Specificity

Anti-FST polyclonal antibody reacts with canine, chicken, bovine, pig, human, mouse, rat, and zebrafish follistatin proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human FST

Application

Anti-FST polyclonal antibody is used to tag follistatin protein for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the possible roles of follistatin in the regulation of TGF-β superfamily mediated processes such as those regulated by myostatin/GDF8 and activin.

Biochem/physiol Actions

Follistatin is a single-chain gonadal protein that specifically inhibits follicle-stimulating hormone release.Follistatin is a single-chain gonadal protein that specifically inhibits follicle-stimulating hormone release. The single FST gene encodes two isoforms, FST317 and FST344 containing 317 and 344 amino acids respectively, resulting from alternative splicing of the precursor mRNA. In a study in which 37 candidate genes were tested for linkage and association with polycystic ovary syndrome (PCOS) or hyperandrogenemia in 150 families, evidence was found for linkage between PCOS and follistatin.

Sequence

Synthetic peptide located within the following region: SLCDELCPDSKSDEPVCASDNATYASECAMKEAACSSGVLLEVKHSGSCN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshimi Oishi et al.
Journal of applied physiology (Bethesda, Md. : 1985), 118(6), 742-749 (2015-01-13)
We examined whether a mixed lactate and caffeine compound (LC) could effectively elicit proliferation and differentiation of satellite cells or activate anabolic signals in skeletal muscles. We cultured C2C12 cells with either lactate or LC for 6 h. We found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service