Skip to Content
Merck
All Photos(1)

Key Documents

AV46789

Sigma-Aldrich

Anti-LMAN2 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C5orf8, Anti-GP36B, Anti-Lectin, mannose-binding 2, Anti-VIP36

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

mouse, dog, rat, human, bovine, horse, rabbit, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LMAN2(10960)

Immunogen

Synthetic peptide directed towards the C terminal region of human LMAN2

Application

Anti-LMAN2 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 0.25μg/mL.

Biochem/physiol Actions

LMAN2 (Lectin, mannose-binding 2) gene encodes a type I transmembrane lectin protein localized in endoplasmic reticulum, the Golgi apparatus and the plasma membrane. It plays a pivotal role in sorting, trafficking and quality control of secretory proteins and/or lipids.[1]

Sequence

Synthetic peptide located within the following region: LMVEHTPDEESIDWTKIEPSVNFLKSPKDNVDDPTGNFRSGPLTGWRVFL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Nawa et al.
Glycobiology, 17(9), 913-921 (2007-06-26)
VIP36 is an intracellular lectin that cycles between the endoplasmic reticulum (ER) and the Golgi apparatus, and is thought to act as a cargo receptor in the transport and sorting of glycoproteins. Here we sought to identify the proteins that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service