Skip to Content
Merck
All Photos(4)

Key Documents

AV40585

Sigma-Aldrich

Anti-HNRPDL antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Heterogeneous Nuclear Ribonucleoprotein D-like

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$643.00

$643.00


Check Cart for Availability


Select a Size

Change View
100 μL
$643.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$643.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

guinea pig, mouse, horse, rat, human, bovine, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNRPDL(9987)

General description

Heterogeneous Nuclear Ribonucleoproteins (hnRNP) are RNA binding proteins that form complexes with heterogeneous nuclear RNA (hnRNA). HnRNPs regulate pre-mRNA processing, metabolism and nuclear cytoplasmic shuttling. Specific HnRNPs have unique nucleic acid binding properties. Heterogeneous Nuclear Ribonucleoprotein D-like (JKTBP) is a DNA- and RNA-binding protein highly expressed in brain and testes tissue. JKTBP interacts with APP (β-amyloid precursor protein) creating a possible link between JKTBP and Alzheimer′s disease.

Specificity

Anti-HNRPDL (JKTBP) polyclonal antibody reacts with canine, zebrafish, human, chicken, rat, bovine, and mouse heterogeneous nuclear ribonucleoprotein D-like proteins.

Immunogen

Synthetic peptide directed towards the middle region of human HNRPDL

Application

Anti-HNRPDL (JKTBP) polyclonal antibody is used to tag the heterogeneous nuclear ribonucleoprotein D-like for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of heterogeneous nuclear ribonucleoprotein D-like in specific mRNA shuttling and Alzheimer′s disease.

Biochem/physiol Actions

HNRPDL belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. HNRPDL has two RRM domains that bind to RNAs.

Sequence

Synthetic peptide located within the following region: TMEDMNEYSNIEEFAEGSKINASKNQQDDGKMFIGGLSWDTSKKDLTEYL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorena Zubović et al.
Nucleic acids research, 40(13), 6255-6269 (2012-03-22)
Mutually exclusive splicing is a form of alternative pre-mRNA processing that consists in the use of only one of a set of two or more exons. We have investigated the mechanisms involved in this process for exon 18 of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service