Skip to Content
Merck
All Photos(1)

Key Documents

AV35300

Sigma-Aldrich

Anti-NMUR2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Neuromedin U receptor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Estimated to ship on29 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NMUR2(56923)

General description

NMUR2 (neuromedin U receptor 2) belongs to the G-protein coupled receptor family. It is known to mediate anti-obesity effects by regulating food intake and body weight.
Rabbit Anti-NMUR2 antibody recognizes human NMUR2.

Immunogen

Synthetic peptide directed towards the N terminal region of human NMUR2

Application

Rabbit Anti-NMUR2 antibody is suitable for western blot applications at a concentration of 0.25 μg/ml.

Biochem/physiol Actions

NMUR2 encodes for one of two G-protein-coupled receptors for the neuropeptide, neuromedin U. This peptide is found in highest levels in the gut and genitourinary system where it potently contracts smooth muscle but is also expressed in the spinal cord and discrete regions of the brain. NMUR2 is highly expressed in the central nervous system.

Sequence

Synthetic peptide located within the following region: MSGMEKLQNASWIYQQKLEDPFQKHLNSTEEYLAFLCGPRRSHFFLPVSV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Peier et al.
Endocrinology, 150(7), 3101-3109 (2009-03-28)
Neuromedin U (NMU) and neuromedin S (NMS) are structurally related neuropeptides that have been reported to modulate energy homeostasis. Pharmacological data have shown that NMU and NMS inhibit food intake when administered centrally and that NMU increases energy expenditure. Additionally

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service