Skip to Content
Merck
All Photos(3)

Key Documents

AV31910

Sigma-Aldrich

Anti-GTF2I antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-General transcription factor II, i

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

110 kDa

species reactivity

rat, rabbit, dog, human, guinea pig, horse, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GTF2I(2969)

General description

GTF2I is a phosphoprotein transcription factor that has six typical repeat motifs. Mutations in this gene have been implicated in Williams Syndrome[1].
Rabbit Anti-GTF2I antibody recognizes chicken, rat, mouse, bovine, human, and canine GTF2I.

Immunogen

Synthetic peptide directed towards the N terminal region of human GTF2I

Application

Rabbit Anti-GTF2I antibody can be used for western blot (0.5μg/ml) and IHC (4-8μg/ml, using paraffin-embedded tissues) assays.

Biochem/physiol Actions

GTF2I encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11.23. The exon(s) encoding 5′ UTR has not been fully defined, but this gene is known to contain at least 34 exons, and its alternative splicing generates 4 transcript variants.

Sequence

Synthetic peptide located within the following region: ILSPGGSCGPIKVKTEPTEDSGISLEMAAVTVKEESEDPDYYQYNIQGSH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colleen A Morris et al.
American journal of medical genetics. Part A, 123A(1), 45-59 (2003-10-14)
Most individuals with Williams syndrome (WS) have a 1.6 Mb deletion in chromosome 7q11.23 that encompasses the elastin (ELN) gene, while most families with autosomal dominant supravalvar aortic stenosis (SVAS) have point mutations in ELN. The overlap of the clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service