Skip to Content
Merck
All Photos(1)

Key Documents

MABS2240

Sigma-Aldrich

Anti-TSH Receptor Antibody, clone CS-17

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$300.00
100 μL
$753.00

$300.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$300.00
100 μL
$753.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

$300.00


Estimated to ship on13 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

CS-17, monoclonal

mol wt

calculated mol wt 86.83 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

agglutination assay: suitable
flow cytometry: suitable
inhibition assay: suitable
radioimmunoassay: suitable

isotype

IgG2aκ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TSHR(7253)

General description

Thyrotropin receptor (UniProt: P16473; also known as Thyroid-stimulating hormone receptor, TSH-R) is encoded by the TSHR (also known as LGR3) gene (Gene ID: 7253) in human. TSH-R is a multi-pass membrane protein of the G-protein coupled receptor (GPCR) family that is synthesized with a signal peptide (aa 1-20), which is subsequently cleaved off to generate the mature form that contains four extracellular domains, seven transmembrane domains, and four cytoplasmic domains. The extracellular domain is linked to a distal signal-specific domain the hinge region that is attached to a transmembrane domain consisting of extracellular and intracellular loops. It serves as a receptor the thyroid-stimulating hormone and TSH binding to this receptor induces thyrocyte growth and proliferation primarily by activating the adenylate cyclase signaling pathway. Clone CS-17 acts allosterically and serves as an inverse agonist for TSH-R constitutive activity. When administered in vivo it is shown to suppress thyroid function and reduces serum T4 levels in BALB/c mice. (Ref.: Chen, RR., et al. (2018). Thyroid. 28(77); 933-940; Chen, CR., et al. (2007). Endocrinology. 148(5); 2375-2382).

Specificity

Clone CS-17 is a mouse monoclonal antibody that detects human Thyrotropin receptor. It targets an epitope within the first extracellular domain from the N-terminal region.

Immunogen

Adenovirus expressing TSH receptor fragment (aa 22-289). adenovirus.

Application

Quality Control Testing

Evaluated by Flow Cytometry in TSH-R transfected HEK293 cells.

Flow Cytometry Analysis: 1 μg of this antibody detected TSH Receptor in one million TSH-R transfected HEK293 cells.

Tested Applications

Flow Cytometry: A representative lot detected TSH Receptor in Flow Cytometry applications (Chen, C.R., et al. (2007). Endocrinology. 148(5):2375-82; Chen, C.R., et al. (2009). Endocrinology. 150(7):3401-8; Chen, C.R., et al. (2018). Thyroid. 28(7):933-940).

Inhibition: A representative lot inhibited the constitutive activity of the TSH Receptor. (Furmaniak, J. et al. (2012). Autoimmun Highlights. 4(1):11-26).

Radioimmunoassay: A representative lot detected TSH Receptor in Radioimmunoassay applications (Chen, C.R., et al. (2007). Endocrinology. 148(5):2375-82).

Antagonist Function: A representative lot bloced TSH binding to the TSH Receptor. Endocrinology. 150(7):3401-8; Chen, C.R., et al. (2018). Thyroid. 28(7):933-940).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TSH Receptor, clone CS-17, Cat. No. MABS2240, is a mouse monoclonal antibody that detects Thyroid-stimulating hormone receptor (TSH-R) and is tested for use in Flow Cytometry, Antagonist Function, Inhibition Assay, and Radioimmunoassay.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service