Skip to Content
Merck
All Photos(1)

Key Documents

AV03050

Sigma-Aldrich

Anti-CDK8 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Cyclin-dependent kinase 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Check Cart for Availability


Select a Size

Change View
100 μL
$547.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

rabbit, bovine, dog, human, mouse, pig, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CDK8(1024)

Immunogen

Synthetic peptide directed towards the C terminal region of human CDK8

Application

Anti-CDK8 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

CDK8 is a regulator of transcription in a multisubunit complex with TFIIH and TEF-β. It exerts positive and negative effects on transcription and is involved signaling that controls nutrient sensing and cell differentiation.[1] CDK8 phosphorylates the regulatory serine residue of STAT1 and mediates JAK-STAT1 signaling.[2]

Sequence

Synthetic peptide located within the following region: GLIMTSDYQRSNPHAAYPNPGPSTSQPQSSMGYSATSQQPPQYSHQTHRY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Staab et al.
JAK-STAT, 2(3), e24275-e24275 (2013-09-27)
Whereas cytokine-induced tyrosine phosphorylation of STAT (signal transducer and activator of transcription) proteins by JAK kinases has been well studied, much less is known about STAT-specific serine kinases and their signal-dependent regulation. The paper by Joanna Bancerek and colleagues published
Josipa Nemet et al.
Biochimie, 97, 22-27 (2013-10-22)
Three cyclin dependent kinases, Cdk7, Cdk8 and Cdk9 are intimately connected with the processes of RNA polymerase II dependent transcription initiation and elongation in eukaryotic cells. Each of these kinases is part of a larger multisubunit complex, TFIIH, Mediator and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service