Skip to Content
Merck
All Photos(1)

Key Documents

A9605

Sigma-Aldrich

AC-93253 iodide

≥98% (HPLC)

Synonym(s):

2-[3-(1,3-Dihydro-1,3,3-trimethyl-2H-indol-2-ylidene)-1-propenyl]-3-ethyl-benzothiazolium iodide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$172.00

$172.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$172.00

About This Item

Empirical Formula (Hill Notation):
C23H25N2SI
CAS Number:
Molecular Weight:
488.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$172.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

dark purple

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

[I-].CC[n+]1c(\C=C\C=C2\N(C)c3ccccc3C2(C)C)sc4ccccc14

InChI

1S/C23H25N2S.HI/c1-5-25-19-13-8-9-14-20(19)26-22(25)16-10-15-21-23(2,3)17-11-6-7-12-18(17)24(21)4;/h6-16H,5H2,1-4H3;1H/q+1;/p-1

InChI key

TWBRXRGFFUATNI-UHFFFAOYSA-M

Biochem/physiol Actions

AC-93253 is a potent, cell permeable, subtype selective RAR (RARα) agonist. EC50 = 6.3 μM. AC-93253 has high selectivity; 89% for RARα vs 67% for RARβ1, 35% for RARβ2, and 11% for RARγ.
Potent, cell permeable, subtype selective RAR (RARα) agonist.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service