Skip to Content
Merck
All Photos(1)

Key Documents

A3236

Sigma-Aldrich

Astaxanthin

≥98% (HPLC)

Synonym(s):

(3S,3′S)-3,3′-Dihydroxy-β,β-carotene-4,4′-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H52O4
CAS Number:
Molecular Weight:
596.84
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic

Assay

≥98% (HPLC)

form

powder

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C2=C(C)C([C@@H](O)CC2(C)C)=O)C(C)(C)C[C@H](O)C1=O

InChI

1S/C40H52O4/c1-27(17-13-19-29(3)21-23-33-31(5)37(43)35(41)25-39(33,7)8)15-11-12-16-28(2)18-14-20-30(4)22-24-34-32(6)38(44)36(42)26-40(34,9)10/h11-24,35-36,41-42H,25-26H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,27-15+,28-16+,29-19+,30-20+/t35-,36-/m0/s1

InChI key

MQZIGYBFDRPAKN-UWFIBFSHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Astaxanthin is a potent carotenoid antioxidant found in marine algae, red yeast and many other plant and animal sources. Animal studies indicate that it reduces blood glucose and ameliorates several parameters of the diabetic metabolic syndrome. It improves blood flow and vascular tone in models of hypertension. In vitro studies indicate that it upregulates connexin 43 and thus, may be chemopreventive against cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Park et al.
Journal of animal science, 91(1), 268-275 (2012-10-27)
Young (2.97±0.01 yr; 8.16±0.15 kg BW) and geriatric (10.71±0.01 yr; 9.46±0.18 kg BW) healthy female Beagle dogs (n=14/age group) were fed 0 or 20 mg astaxanthin daily for 16 wk to examine modulation of mitochondrial function. Fasted blood was sampled
Takehiro Kiko et al.
PloS one, 7(11), e49620-e49620 (2012-11-21)
Amyloid β-peptide (Aβ) is hypothesized to play a key role by oxidatively impairing the capacity of red blood cells (RBCs) to deliver oxygen to the brain. These processes are implicated in the pathogenesis of Alzheimer's disease (AD). Although plasma Aβ
Marina Wayama et al.
PloS one, 8(1), e53618-e53618 (2013-01-18)
Haematococcus pluvialis is a freshwater species of green algae and is well known for its accumulation of the strong antioxidant astaxanthin, which is used in aquaculture, various pharmaceuticals, and cosmetics. High levels of astaxanthin are present in cysts, which rapidly
R T Lorenz et al.
Trends in biotechnology, 18(4), 160-167 (2000-03-31)
As a result of high production costs, commercial products from microalgae must command high prices. Astaxanthin produced by Haematococcus is a product that has become a commercial reality through novel and advanced technology. Cultivation methods have been developed to produce
P F Zagalsky
Acta crystallographica. Section D, Biological crystallography, 59(Pt 8), 1529-1531 (2003-07-24)
The crystal structure of a beta-crustacyanin allows an analysis of the various proposals for the mechanism of the bathochromic shift from orange to purple-blue of astaxanthin in this lobster carotenoprotein. Structural and previous chemical and biophysical studies suggest that extension

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service