Skip to Content
Merck
All Photos(1)

Documents

11775

Sigma-Aldrich

Canthaxanthin

≥95.0% (HPLC), powder or crystals

Synonym(s):

Canthaxanthin (trans), β-Carotin-4,4′-dione, E 161g

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H52O2
CAS Number:
Molecular Weight:
564.84
Beilstein:
1898520
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

product name

Canthaxanthin, ≥95.0% (HPLC)

biological source

synthetic

Assay

≥95.0% (HPLC)

form

powder or crystals

color

light red to dark brown

λ

in hexane (with 2% dichloromethane)

UV absorption

λ: 465 nm±5 nm Amax

storage temp.

−20°C

SMILES string

CC(=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C1=C(C)C(=O)CCC1(C)C)\C=C\C=C(C)\C=C\C2=C(C)C(=O)CCC2(C)C

InChI

1S/C40H52O2/c1-29(17-13-19-31(3)21-23-35-33(5)37(41)25-27-39(35,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-36-34(6)38(42)26-28-40(36,9)10/h11-24H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+

InChI key

FDSDTBUPSURDBL-DKLMTRRASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Canthaxanthin (red diketocarotenoid or 4, 4′-diketo-β-carotene), is a common xanthophyll found naturally in microorganisms and marine organisms, as well as in certain animals. It acts as an antioxidant in living organisms. The potential antioxidant activity of canthaxanthin is due to the presence of conjugated double bonds in its structure. It finds applications in various fields including poultry, fishery, cosmetics, medicine, and pharmaceuticals. In food industries, canthaxanthin can be used as a coloring agent.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Canthaxanthin and excess vitamin A alter α -tocopherol, carotenoid and iron status in adult rats
Blakely SR, et al.
The Journal of Nutrition, 121(10), 1649-1655 (1991)
Astaxanthin and canthaxanthin are potent antioxidants in a membrane model
Palozza P and Krinsky NI
Archives of Biochemistry and Biophysics, 297(2), 291-295 (1992)
Efficient syntheses of the keto-carotenoids canthaxanthin, astaxanthin, and astacene
Choi, Seyoung and Koo, Sangho
The Journal of Organic Chemistry, 70(8), 3328-3331 (2005)
Use of response surface methodology in a fed-batch process for optimization of tricarboxylic acid cycle intermediates to achieve high levels of canthaxanthin from Dietzia natronolimnaea HS-1
Nasrabadi MRN and Razavi SH
Journal of Bioscience and Bioengineering, 109(4), 361-368 (2010)
Canthaxanthin
Gupta AK, et al.
International Journal of Dermatology, 24(1), 528-532 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service